Thermo Scientific Chemicals

Cadmium selenide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 036323.22
100 g, Each
Thermo Scientific Chemicals

Cadmium selenide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 036323.22
100 g, Each
Quantity
Catalog number: 036323.22
also known as 036323-22
Price (USD)

Chemical Identifiers

CAS
1306-24-7
IUPAC Name
selanylidenecadmium
Molecular Formula
CdSe
InChI Key
AQCDIIAORKRFCD-UHFFFAOYSA-N
SMILES
[Se]=[Cd]
Total Metal Impurities
0.005% max.

Description

It ia a photoconductive and semiconductor material used in a cell where a fast response time and high sensitivity to longer wavelengths of light is required; also used as a red pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text