Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%
Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%
Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%
Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%
Thermo Scientific Chemicals

Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%

CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 036375.30
also known as 036375-30
Price (USD)
614.65
Special offer
Online exclusive
Ends: 26-Sep-2025
759.00
Save 144.35 (19%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
614.65
Special offer
Online exclusive
Ends: 26-Sep-2025
759.00
Save 144.35 (19%)
Each
Add to cart
Specifications
Assay Percent Notes(metals basis excluding Ni), Ni nominal 1%
CAS7440-48-4
ChEBICHEBI:27638
Chemical Name or MaterialCobalt pieces
ColorGray
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only