Copper foil, 0.25mm (0.01in) thick, annealed, 99.95% (metals basis), oxygen free
Copper foil, 0.25mm (0.01in) thick, annealed, 99.95% (metals basis), oxygen free
Copper foil, 0.25mm (0.01in) thick, annealed, 99.95% (metals basis), oxygen free
Copper foil, 0.25mm (0.01in) thick, annealed, 99.95% (metals basis), oxygen free
Thermo Scientific Chemicals

Copper foil, 0.25mm (0.01in) thick, annealed, 99.95% (metals basis), oxygen free

Form: Approx. 5.6g/50 x 50mm | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 250 mm
50 x 50 mm
Catalog number 036396.GB
also known as 036396-GB
Price (USD)
221.65
Special offer
Online exclusive
260.00
Save 38.35 (15%)
Each
Add to cart
Quantity:
50 x 250 mm
Request bulk or custom format
Price (USD)
221.65
Special offer
Online exclusive
260.00
Save 38.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Copper
Total Metal Impurities0.05% max.
Comment0.23 - 0.28mm thick
FormFoil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only