Thermo Scientific Chemicals

D-Sorbitol, 98%

Catalog number: 036404.A3
2 kg, Each
Thermo Scientific Chemicals

D-Sorbitol, 98%

Catalog number: 036404.A3
2 kg, Each
Quantity
Catalog number: 036404.A3
also known as 036404-A3
Price (USD)
Quantity
-

Chemical Identifiers

CAS
50-70-4
IUPAC Name
hexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H14O6
InChI Key
FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)C(O)CO
Appearance (Color)
White
Form
Powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Residual water
≤1.5%
Residue on ignition
≤0.1%

Description

D-sorbitol is added to food products as sweetener. It is also used in industrial and medical applications. Sorbitol often is applied in modern cosmetics as a humectant and thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text