Tannic acid, ACS
Tannic acid, ACS
Tannic acid, ACS
Thermo Scientific Chemicals

Tannic acid, ACS

CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 036410.22
also known as 036410-22
Price (EUR)
48,50
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
48,50
Each
Add to cart
Chemical Identifiers
CAS1401-55-4
IUPAC Name2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate
Molecular FormulaC76H52O46
InChI KeyLRBQNJMCXXYXIU-UHFFFAOYNA-N
SMILESOC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentDextrin P.T.
Impurity contentIdentification P.T.
Impurity contentSugars P.T.
Impurity contentZn 0.005%
Loss on Drying12.0%
View more
Tannic acid has anti-bacterial, anti-enzymatic and astringent applications. Used in the dyeing process for cellulose fibers such as cotton. Used in food and medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only