Gold Nickel wire, 0.5mm (0.02in) dia, annealed, 99.85% (metals basis)
Thermo Scientific Chemicals

Gold Nickel wire, 0.5mm (0.02in) dia, annealed, 99.85% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
1 m
Catalog number 036451.BS
also known as 036451-BS
Price (USD)
200.00
Each
Quantity:
10 cm
Request bulk or custom format
Price (USD)
200.00
Each
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeAu:Ni; 82:18 wt%
Chemical Name or MaterialGold Nickel wire
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only