Thermo Scientific Chemicals

Lecithin, Refined

Catalog number: 036486.A1
1 kg, Each
Thermo Scientific Chemicals

Lecithin, Refined

Catalog number: 036486.A1
1 kg, Each
Quantity
Catalog number: 036486.A1
also known as 036486-A1
Price (USD)
Price: 167.00
Online price: 141.65
Your price:
Quantity
-

Chemical Identifiers

CAS
8002-43-5
IUPAC Name
(2-{[3-(hexadecanoyloxy)-2-(octadeca-9,12-dienoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular Formula
C42H80NO8P
InChI Key
JLPULHDHAOZNQI-UHFFFAOYNA-N
SMILES
CCCCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Assay (unspecified)
Hexane insoluble 0.3%, max
Acid Value
<35 mg KOH/g
Residual water
< 1%
Peroxide content
5 meq /kg, max
Appearance (Color)
Light tan to medium yellow

Description

Lecithin is used as an emulsifying and stabilizing agent in the food, pharmaceutical, and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacturing of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text