Thermo Scientific Chemicals

Dimethyl sulfoxide-d{6}, 99.9% (Isotopic), Thermo Scientific Chemicals

Catalog number: 036517.48
10 x 1 g, Each
Thermo Scientific Chemicals

Dimethyl sulfoxide-d{6}, 99.9% (Isotopic), Thermo Scientific Chemicals

Catalog number: 036517.48
10 x 1 g, Each
Quantity
Catalog number: 036517.48
also known as 036517-48
Price (USD)

Chemical Identifiers

CAS
2206-27-1
IUPAC Name
(²H₃)methanesulfinyl(²H₃)methane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Appearance (Color)
Clear, colorless
Assay by NMR
99.9% min., Isotopic enrichment
Form
liquid

Description

Deuterated DMSO is a common solvent used in NMR spectroscopy. It is in combination with pyridine-d5 plays an important role for solution-state 2D NMR of ball-milled plant cell wall gel. Low-energy collisions of ionized acetone-d6, DMSO-d6 and pyridine-d5 with alkanethiolate is used to prepare self-assembled monolayer surfaces of gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text