Zinc granules, ACS, -20 mesh, 99.8% min
Zinc granules, ACS, -20 mesh, 99.8% min
Zinc granules, ACS, -20 mesh, 99.8% min
Thermo Scientific Chemicals

Zinc granules, ACS, -20 mesh, 99.8% min

Zinc Granules; ACS; 99.8%; Alfa Aesar; -20 Mesh | CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number 036602.22
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.50
Save 6.85 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.50
Save 6.85 (15%)
Each
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentFe: ≤0.01%
Impurity contentPb: ≤ 0.01%
Particle Size20 mesh
Appearance (Color)Silver-gray
FormGranules
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only