Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Thermo Scientific Chemicals

Formic acid, ACS, 96+%

CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
2 kg
4 x 500 g
產品號碼 036617.A3
亦稱為 036617-A3
價格 (TWD)
-
Quantity:
2 kg
Request bulk or custom format
化學識別
CAS64-18-6
IUPAC Nameformic acid
Molecular FormulaCH2O2
InChI KeyBDAGIHXWWSANSR-UHFFFAOYSA-N
SMILESOC=O
檢視更多
規格Specification Sheet規格表
Chloride content0.001%, max
Heavy Metals0.001%, max ( as Pb)
Color (APHA)15, max
Impurity contentSO₄ 0.003%, max
CommentDilution Test P.T.
檢視更多
Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only