Thermo Scientific Chemicals

2-Chloroethanol, 99%, Thermo Scientific Chemicals

Catalog number: 036636.AK
250 mL, Each
Thermo Scientific Chemicals

2-Chloroethanol, 99%, Thermo Scientific Chemicals

Catalog number: 036636.AK
250 mL, Each
Quantity

Chemical Identifiers

CAS
107-07-3
IUPAC Name
2-chloroethan-1-ol
Molecular Formula
C2H5ClO
InChI Key
SZIFAVKTNFCBPC-UHFFFAOYSA-N
SMILES
OCCCl
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Form
Liquid

Description

2-Chloroethanol is used in the preparation of several dyes by the alkylation of aniline derivatives with chloroethanol. It is a building block in the production of pharmaceuticals and plasticizers. Also used for manufacture of thiodiglycol. It is a solvent for cellulose acetate and ethyl cellulose, textile printing dyes, in dewaxing, refining of rosin, extraction of pine lignin and the cleaning of machines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text