Thermo Scientific Chemicals

Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals

1L, Each
Thermo Scientific Chemicals

Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals

1L, Each
Quantity
Catalog number: 036645.K2
also known as 036645-K2
Price (USD)
Price: 34.30
Your Price:
Quantity
-

Chemical Identifiers

CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
Conductivity
0.000002 Siemens per cm @ 25?C max
Impurity content
Silicate (as SiO₂) : 0.01ppm max
Chloride content
0.4ppm max
Impurity content
(Nitrate, NO₃) : 0.4ppm max
Impurity content
(Sulphate, SO₄) : 1ppm

Description

Used in conductometric titration, Synthesis of nanomaterials. Mostly it is used as solvent in industries and in all research fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only