4-Dimethylaminobenzaldehyde, ACS
4-Dimethylaminobenzaldehyde, ACS
4-Dimethylaminobenzaldehyde, ACS
Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, ACS

CAS: 100-10-7 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 036684.14
also known as 036684-14
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.00
Save 5.35 (13%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.00
Save 5.35 (13%)
Each
Chemical Identifiers
CAS100-10-7
IUPAC Name4-(dimethylamino)benzaldehyde
Molecular FormulaC9H11NO
InChI KeyBGNGWHSBYQYVRX-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.1%
Solution Testin alcohol P.T.
Appearance (Color)Color of Hydrochloric acid solution: P.T.
Melting Point73 TO 75 Deg C
Color (APHA)Of alcohol solution: 60, max
View more
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only