Thermo Scientific Chemicals

Molybdenum(VI) oxide, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 036687.22
100 g, Each
Thermo Scientific Chemicals

Molybdenum(VI) oxide, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 036687.22
100 g, Each
Quantity
Catalog number: 036687.22
also known as 036687-22
Price (USD)
Price: 71.90
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1313-27-5
IUPAC Name
trioxomolybdenum
Molecular Formula
MoO3
InChI Key
JKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILES
O=[Mo](=O)=O
Assay (unspecified)
99.5% MoO₃ min
Impurity content
Arsenate, phosphate, and silicate (as SiO₂) 0.001%
Impurity content
NO₃ P.T. (limit about 0.003%)
Impurity content
PO₄ 5ppm
Trace Metals
(as Pb) 0.005%

Description

Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in oil industr. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Source of molybdenum compounds, in agriculture, in analytical chemistry, as a corrosion inhibitor, in ceramic glazes, enamels and pigments, as a catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text