Cyclohexyldiphenylphosphine, 98%
Cyclohexyldiphenylphosphine, 98%
Cyclohexyldiphenylphosphine, 98%
Thermo Scientific Chemicals

Cyclohexyldiphenylphosphine, 98%

CAS: 6372-42-5 | C18H21P | 268.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
100 g
Catalog number 036708.03
also known as 036708-03
Price (USD)
35.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.80
Each
Chemical Identifiers
CAS6372-42-5
IUPAC Namecyclohexyldiphenylphosphane
Molecular FormulaC18H21P
InChI KeyZXKWUYWWVSKKQZ-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder or solid
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (dry wt. basis) (non-U.S. specification)
Assay by NMR≥97.5% (31P NMR: U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Cyclohexyldiphenylphosphine is used as ligand in coupling reaction. It is also used as a medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only