Ground & Waste Water Pollution Standard Solution 3, Specpure™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ground & Waste Water Pollution Standard Solution 3, Specpure™, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
036750.AE
also known as 036750-AE
100 mL
Catalog number 036750.AE
also known as 036750-AE
Price (EUR)
330,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
330,00
Each
Specifications
Chemical Name or MaterialGround & Waste Water Pollution Standard Solution 3
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only