Bis(carboxyethylgermanium) sesquioxide, 99.7%
Bis(carboxyethylgermanium) sesquioxide, 99.7%
Bis(carboxyethylgermanium) sesquioxide, 99.7%
Thermo Scientific Chemicals

Bis(carboxyethylgermanium) sesquioxide, 99.7%

CAS: 12758-40-6 | C6H10Ge2O7 | 339.399 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 036758.06
also known as 036758-06
Price (EUR)
78,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
78,90
Each
Specifications
CAS12758-40-6
ChEBICHEBI:32060
Chemical Name or MaterialBis(carboxyethylgermanium) sesquioxide
EINECS Number235-800-0
Formula Weight339.25
View more
A bifunctional metalloligand with metal-C (Ge-C) bonds,bis(carboxyethylgermanium) sesquioxide, has been used to make two metal-organometallic frameworks (MOMFs). bis-(carboxyethylgermanium)sesquioxide is a as a promising anti-oxidative agent. Bis(carboxyethylgermanium) sesquioxide induce INF-γ and has undergone clinical trials with promising preclinical results.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only