Dionex™ Inorganic Anion and Oxyhalide Standards
Thermo Scientific™

Dionex™ Inorganic Anion and Oxyhalide Standards

Spend less time preparing standards and more time generating ion chromatography results with Dionex Ion Standards for routine anion and oxyhalide analyses.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
037160Sulfate Standard
303167Chlorite Standard
303168Bromate Standard
303169Nitrite Standard
303170Chlorate Standard
303172Phosphate Standard
303173Nine Anion Standard
056497Nitrate Standard
037157Combined Five Anion Standard
037158Fluoride Standard
037159Chloride Standard
057590Combined Seven Anion Standard II
Catalog number 037160
Price (USD)
140.00
Each
Description:
Sulfate Standard
Price (USD)
140.00
Each

Increase your productivity and reduce standard preparation errors with Thermo Scientific™ Dionex™ inorganic anion and oxyhalide standards for ion chromatography. Available for routine anion and oxyhalide determinations, these single and combined ion standards are made with high purity, reagent-grade chemicals and water. Each standard is NIST traceable and includes a Certificate of Analysis.

Increase Productivity
  • Easy and convenient IC standard preparation
  • Reduce standard preparation time
  • Reduce time-consuming standard validation
Guarantee the Reliability of Your Analyses
  • Reduce standard preparation errors
  • Increase accuracy of quantifications
  • Improve day-to-day reproducibility
The Quality You Expect from Thermo Fisher Scientific
  • All standards are made from high-purity, reagent-grade chemicals and water (18 MΩ-cm)
  • Single ion standards are checked for ionic contamination
  • All standards are National Institute of Standards and Technology (NIST) traceable
  • A Certificate of Analysis is provided with every standard
Unconditional Guarantee

Your satisfaction is guaranteed. All Thermo Scientific Dionex standards carry a 90-day warranty.

Specifications
Concentration1000 mg/L
DescriptionSulfate Standard
For Use With (Application)HPLC
HazardsEyes/Mucous Membrane Irritant
OperationUsed as a standard solution.
Quantity100 mL
Standard ComponentsDisodium Sulfate Aqueous Solution
Standard Concentration1000 mg/L
StorageRoom Temperature
Volume (Metric)100 mL
TypeStandard
Unit SizeEach