Dionex™ 0.5M Sodium Carbonate Anion Eluent Concentrate
Thermo Scientific™
Dionex™ 0.5M Sodium Carbonate Anion Eluent Concentrate
For fast, reliable anion eluent preparation, this ready-to-dilute eluent concentration saves time and eliminates potential contamination or preparation error.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripción
037162Concentrado de carbonato sódico, 0,5 M;
037163Concentrado de bicarbonato sódico
Número de catálogo 037162
Precio (MXN)
-
Descripción:
Concentrado de carbonato sódico, 0,5 M;
For fast, reliable anion eluent preparation, the ready-to-dilute Thermo Scientific™ Dionex™ 0.5M Carbonate Eluent Concentrate will save you time and eliminate potential contamination or preparation error. The 500mL of 0.5M carbonate eluent concentrate is quality checked for ionic contamination, made with only high purity, reagent-grade chemicals and water (18 MO-cm). Use this concentrate in conjunction with the Dionex 0.5M Bicarbonate Eluent Concentrate for custom eluent concentrations.
  • Only high-purity, reagent-grade chemicals and water (18 MΩ-cm), quality-control checked for ionic contamination, are used
  • All standards are NIST™ traceable
Includes:
A Certificate of Analysis is provided with every standard and chelation chromatography reagent
Especificaciones
Certificaciones/conformidadTrazable al NIST
Concentración0,5 M
DescripciónConcentrado de carbonato sódico, 0,5 M;
Para utilizar con (equipo)Columnas IonPac AS4A, AS4A-SC y AS9-SC
RiesgosIrritante
IncluyeCertificado de análisis
Cantidad500 mL
Componentes estándarSolución acuosa de carbonato sódico
Concentración estándar0,5 M
AlmacenamientoTemperatura ambiente
TipoConcentrado de eluyentes
Unit SizeEach 1