Platinum, 3% on activated carbon powder, standard, reduced, nominally 50% water wet
Thermo Scientific Chemicals

Platinum, 3% on activated carbon powder, standard, reduced, nominally 50% water wet

Form: 900 - 1000m2 Standard, reduced, nominally 50% water wet | CAS: 7440-6-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 038316.06
also known as 038316-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
AppearanceBlack powder
CAS6-6-7440
Chemical Name or MaterialPlatinum
ColorBlack
Health Hazard 1H228
View more
Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jewellery. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only