Platinum, 1% on granular carbon, reduced, nominally 50% water wet
Platinum, 1% on granular carbon, reduced, nominally 50% water wet
Platinum, 1% on granular carbon, reduced, nominally 50% water wet
Thermo Scientific Chemicals

Platinum, 1% on granular carbon, reduced, nominally 50% water wet

CAS: 7440-6-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 038343.22
also known as 038343-22
Price (TWD)
-
Quantity:
100 g
Request bulk or custom format
Specifications
CAS6-6-7440
Chemical Name or MaterialPlatinum
ColorGray
Health Hazard 1H228
Health Hazard 3P210-P240-P241-P280-P370+P378q
View more
Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jeweler. It also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells. As catalyst for curing of plastics, as catalyst for dehydrogenation, isomerization, and oxidation in the production of sulfuric, nitric, and organic acids and amines, for surgical instruments and implants. Platinum nanoparticles are used in scanning probes (microscopy) and catalysts. Catalyst used for the hydrogenation of alkenes, alkynes, nitro groups, ketones, aromatics, used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols. Schiff's base and nitrohydrogenation, hydrosilation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only