Phenol Red sodium salt, 0.02% w/v aq. soln.
Phenol Red sodium salt, 0.02% w/v aq. soln.
Phenol Red sodium salt, 0.02% w/v aq. soln.
Thermo Scientific Chemicals

Phenol Red sodium salt, 0.02% w/v aq. soln.

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
数量:
100 mL
500 mL
製品番号(カタログ番号) 038702.AE
または、製品番号038702-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
仕様
CAS34487-61-1
化学物質名または材質Phenol Red sodium salt
濃度0.02% w/v aq. soln.
密度0.972 g/mL
EINECS番号252-057-8
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only