Phenolphthalein, 0.5% w/v in alcohol
Phenolphthalein, 0.5% w/v in alcohol
Phenolphthalein, 0.5% w/v in alcohol
Phenolphthalein, 0.5% w/v in alcohol
Thermo Scientific Chemicals

Phenolphthalein, 0.5% w/v in alcohol

CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 038703.K2
also known as 038703-K2
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS77-09-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC20H14O4
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Concentration (w/v)0.5% w/v in alcohol
Transition IntervalpH 8 colorless to pH 10 red
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use