Thermo Scientific Chemicals

Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Catalog number: 038714.K2
1 L, Each
Thermo Scientific Chemicals

Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Catalog number: 038714.K2
1 L, Each
Quantity
Catalog number: 038714.K2
also known as 038714-K2
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
1336-21-6
Form
Liquid
Assay from Supplier's CofA
pH 4.01 (± 0.022 @ 25°C)
Appearance (Color)
Clear colorless

Description

Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text