Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL
Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL
Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL
Thermo Scientific Chemicals

Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
產品號碼 038726.AE
亦稱為 038726-AE
價格 (TWD)
-
Quantity:
100 mL
Request bulk or custom format
規格
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum
Health Hazard 1H315-H319
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only