Thermo Scientific Chemicals

Glycerol, ultrapure, HPLC Grade, Thermo Scientific Chemicals

4 x 1 L, Each
Thermo Scientific Chemicals

Glycerol, ultrapure, HPLC Grade, Thermo Scientific Chemicals

4 x 1 L, Each
Quantity
Catalog number: 038988.M1
also known as 038988-M1
Price (USD)
Price: 425.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
56-81-5
IUPAC Name
propane-1,2,3-triol
Molecular Formula
C3H8O3
InChI Key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES
OCC(O)CO
Assay (GC)
99.0%, min
Optical Absorbance at
206 nm 1.00 AU, max
Optical Absorbance at
225 nm 0.25 AU, max
Optical Absorbance at
320 nm 0.02 AU, max
Optical Absorbance at
210 nm 0.70 AU, max

Description

Glycerol is used as emulsifier, thickening agent in cosmetics and foods, plasticizer in packaging, paints and paper industry, preservative in food and beverages. It is used in pharmaceutical preparations to improve smoothness and providing lubrication. It is also used in epichlorohydrin and mono propylene glycol production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only