Thermo Scientific Chemicals

Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals

4 L, Each
Thermo Scientific Chemicals

Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals

4 L, Each
Quantity
Catalog number: 038994.K7
also known as 038994-K7
Price (USD)
Price: 222.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
109-99-9
IUPAC Name
oxolane
Molecular Formula
C4H8O
InChI Key
WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES
C1CCOC1
Form
Liquid
Comment
Sub-micron filtered
Peroxide content
0.015%, max ( at time of packaging)
Stabilizer
0.02-0.03% BHT
Appearance (Color)
Colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only