Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT
Thermo Scientific Chemicals

Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
Catalog number 038994.M1
also known as 038994-M1
Price (EUR)
277,00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (EUR)
277,00
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Peroxide content0.015%, max ( at time of packaging)
Stabilizer0.02-0.03% BHT
Assay (GC)≥99.7%
Water Content (Karl Fischer Titration)0.03%, max
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only