1,2-Dichloroethane, ACS, 99+%
1,2-Dichloroethane, ACS, 99+%
1,2-Dichloroethane, ACS, 99+%
Thermo Scientific Chemicals

1,2-Dichloroethane, ACS, 99+%

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, H2O 0.03% | CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039121.K7
also known as 039121-K7
Price (USD)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay from Suppliers CofA≥99.0%
Free acid (titration)≤0.0003 meq/g
View more
Used to make vinyl chloride, and chlorinated solvents(trichloroethane, trichloroethylene, perchloroethylene, and vinylidene), remove grease, resins, glue and dirt. Used as a solvent in the manufacturing of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. Used as fumigant residues for wheat, flour, bran, middlings and bread. Used as solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only