Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol, Thermo Scientific Chemicals
Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number 039154.AE
also known as 039154-AE
Price (USD)/ Each
241.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
241.00
Add to cart
Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol, Thermo Scientific Chemicals
Catalog number039154.AE
Price (USD)/ Each
241.00
-
Add to cart
Specifications
Chemical Name or MaterialLithium niobium methoxide
CAS21864-11-9
Health Hazard 1H225-H301+H311+H331-H314-H335-H370
Health Hazard 2GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Used as a precursor for chemical vapor deposition of thin films for lithium ion battery electrolyte materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only