Cerium(III) phosphate, 99% min
Cerium(III) phosphate, 99% min
Cerium(III) phosphate, 99% min
Thermo Scientific Chemicals

Cerium(III) phosphate, 99% min

CAS: 13454-71-2 | CeO4P | 235.09 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
產品號碼 039215.14
亦稱為 039215-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS13454-71-2
IUPAC Namecerium(3+) phosphate
Molecular FormulaCeO4P
InChI KeyTYAVIWGEVOBWDZ-UHFFFAOYSA-K
SMILES[Ce+3].[O-]P([O-])([O-])=O
檢視更多
規格Specification Sheet規格表
Assay (unspecified)98.6% min
Intermediate product in cerium-based technique for staining lysosomal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only