Bis(pentamethylcyclopentadienyl)nickel
Bis(pentamethylcyclopentadienyl)nickel
Bis(pentamethylcyclopentadienyl)nickel
Thermo Scientific Chemicals

Bis(pentamethylcyclopentadienyl)nickel

CAS: 74507-63-4 | C20H30Ni | 329.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 039238.06
also known as 039238-06
Price (USD)
790.65
Online Exclusive
878.00
Save 87.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
790.65
Online Exclusive
878.00
Save 87.35 (10%)
Each
Specifications
CAS74507-63-4
Chemical Name or MaterialBis(pentamethylcyclopentadienyl)nickel
Formula Weight329.17
Health Hazard 1H350
Health Hazard 2GHS H Statement
H350
May cause cancer.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only