Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Thermo Scientific Chemicals

Iron(III) 1,1,1-trifluoro 2,4-pentanedionate

CAS: 14526-22-8 | C15H12F9FeO6 | 515.09 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 039265.06
亦稱為 039265-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
規格
CAS14526-22-8
Chemical Name or MaterialIron(III) 1,1,1-trifluoro 2,4-pentanedionate
Formula Weight515.09
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
檢視更多
It is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only