Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%
Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%
Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%
Thermo Scientific Chemicals

Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%

CAS: 14874-82-9 | C7H7O4Rh | 258.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 039295.MD
also known as 039295-MD
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS14874-82-9
IUPAC Nameλ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
Molecular FormulaC7H7O4Rh
InChI KeyBZCAWKOPWNIDOC-FGSKAQBVSA-M
SMILES[Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≤96.0%
Appearance (Color)Green
FormCrystalline powder or powder
(Acetylacetonato)dicarbonylrhodium(I) is used in in situ formation of a floors-soluble hydroformylation catalyst of interest in molecular engineering. It is also used as a catalyst for various carbonylation reactions, silylcarbocyclizations, conjugate additions to eons, carbamoylstannation, and reduction of aromatic nitro compounds. It is also used in Hydroformylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only