Bis(cyclopentadienyl)molybdenum dichloride, 99%
Bis(cyclopentadienyl)molybdenum dichloride, 99%
Bis(cyclopentadienyl)molybdenum dichloride, 99%
Thermo Scientific Chemicals

Bis(cyclopentadienyl)molybdenum dichloride, 99%

CAS: 12184-22-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 039393.04
also known as 039393-04
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Specifications
CAS12184-22-4
Chemical Name or MaterialBis(cyclopentadienyl)molybdenum dichloride
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight297.04
Health Hazard 1H314-H335
View more

Bis(cyclopentadienyl)molybdenum dichloride, 99% is used to prepare many derivatives. It is also used as pharmaceutical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only