Potassium selenite
Potassium selenite
Potassium selenite
Thermo Scientific Chemicals

Potassium selenite

CAS: 10431-47-7 | K2O3Se | 205.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 039436.14
also known as 039436-14
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS10431-47-7
IUPAC Namedipotassium selenite
Molecular FormulaK2O3Se
InChI KeyRNGFNLJMTFPHBS-UHFFFAOYSA-L
SMILES[K+].[K+].[O-][Se]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Se 38% min
One among the compounds used in cattle feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only