Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate
Thermo Scientific Chemicals

Nickel(II) oxalate dihydrate

CAS: 6018-94-6 | C2H4NiO6 | 182.741 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 039454.18
also known as 039454-18
Price (EUR)
92,60
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
92,60
Each
Specifications
CAS6018-94-6
Chemical Name or MaterialNickel(II) oxalate dihydrate
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECS Number208-933-7
Formula Weight182.76 (146.73 Anhydrous)
View more
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only