Bis(cyclopentadienyl)zirconium dichloride, 99+%, Thermo Scientific Chemicals
Bis(cyclopentadienyl)zirconium dichloride, 99+%, Thermo Scientific Chemicals
Bis(cyclopentadienyl)zirconium dichloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(cyclopentadienyl)zirconium dichloride, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039558.14
also known as 039558-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1291-32-3
IUPAC Namedichlorobis(cyclopenta-2,4-dien-1-yl)zirconium
Molecular FormulaC10H10Cl2Zr
InChI KeyQRUYYSPCOGSZGQ-UHFFFAOYSA-L
SMILESCl[Zr](Cl)(C1C=CC=C1)C1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormCrystalline powder
Assay (Titration)> 99.0%
Zirconocene dichloride can also be used to prepare the Negishi reagent, Cp2Zr(η2-butene) which is used as a source for Cp2Zr in oxidative cyclisation reactions. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only