Dimethylzinc, 96%
Dimethylzinc, 96%
Dimethylzinc, 96%
Thermo Scientific Chemicals

Dimethylzinc, 96%

CAS: 544-97-8 | C2H6Zn | 95.45 g/mol
Have Questions?
Catalog NumberQuantityPackaging
039563.14
also known as 039563-14
25 gCylinder
Catalog number 039563.14
also known as 039563-14
Price (USD)
1,130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,330.00
Save 199.35 (15%)
Each
Quantity:
25 g
Packaging:
Cylinder
Request bulk or custom format
Price (USD)
1,130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,330.00
Save 199.35 (15%)
Each
Chemical Identifiers
CAS544-97-8
IUPAC Namedimethylzinc
Molecular FormulaC2H6Zn
InChI KeyJRPGMCRJPQJYPE-UHFFFAOYSA-N
SMILES[CH3-].[CH3-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95.1% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only