Diphenyltin oxide, Thermo Scientific Chemicals
Diphenyltin oxide, Thermo Scientific Chemicals
Diphenyltin oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenyltin oxide, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
039615.14
also known as 039615-14
25 g
Catalog number 039615.14
also known as 039615-14
Price (USD)
238.65
Online Exclusive
265.00
Save 26.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
238.65
Online Exclusive
265.00
Save 26.35 (10%)
Each
Specifications
CAS2273-51-0
Chemical Name or MaterialDiphenyltin oxide
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECS Number218-883-8
Formula Weight288.9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only