Silver molybdenum oxide, 99%
Silver molybdenum oxide, 99%
Silver molybdenum oxide, 99%
Thermo Scientific Chemicals

Silver molybdenum oxide, 99%

CAS: 13765-74-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 039650.01
also known as 039650-01
Price (TWD)
-
Quantity:
0.25 g
Request bulk or custom format
Chemical Identifiers
CAS13765-74-7
IUPAC Namedisilver(1+) dioxomolybdenumbis(olate)
Molecular FormulaAg2MoO4
InChI KeyMHLYOTJKDAAHGI-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only