Tin oxide, antimony doped, polymeric precursor, Oxide ≈ 30 wt%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin oxide, antimony doped, polymeric precursor, Oxide ≈ 30 wt%, Thermo Scientific Chemicals

Used in transparent electrical conducting electrodes
Catalog NumberQuantity
039754.14
also known as 039754-14
25 g
Catalog number 039754.14
also known as 039754-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Specifications
CAS1332-29-2
Chemical Name or MaterialTin oxide
Concentration or Composition (by Analyte or Components)Oxide ≈30 wt%
Density1.61 g/mL
Health Hazard 1Warning
View more
Used in transparent electrical conducting electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only