Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution
Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution
Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution

CAS: 139-33-3 | C10H14N2Na2O8 | 336.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 039783.K7
also known as 039783-K7
Price (USD)
179.00
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
179.00
Each
Chemical Identifiers
CAS139-33-3
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular FormulaC10H14N2Na2O8
InChI KeyZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Concentration0.200N ± 0.19% (0.0019N)
Ethylenediaminetetraacetic acid disodium salt is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics, it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only