Thermo Scientific Chemicals

Carbon disulfide, ACS, 99.9+%, Thermo Scientific Chemicals

Catalog number: 039785.K3
2 L, Each
Thermo Scientific Chemicals

Carbon disulfide, ACS, 99.9+%, Thermo Scientific Chemicals

Catalog number: 039785.K3
2 L, Each
Quantity
Catalog number: 039785.K3
also known as 039785-K3
Price (USD)
Price: 200.00
Online price: 170.65
Your price:
Quantity
-

Chemical Identifiers

CAS
75-15-0
IUPAC Name
methanedithione
Molecular Formula
CS2
InChI Key
QGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILES
S=C=S
Color (APHA)
≤10
Assay (GC)
≤99.9%
Residual water
≤0.05%
Impurity content
Hydrogen sulfide (H₂S) P.T. (limit about 1.5ppm)
Residue on Evaporation
≤0.002%

Description

It is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reaction agent for the production of rubber vulcanization. accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Evaporation Residue 0.002%, Hydrogen sulfide (H2S) P.T. (limit about 1.5ppm, Sulfur dioxide (SO2) P.T. (limit about 2.5ppm), H2O 0.05%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text