Product Image
Thermo Scientific™
Dionex™ OnGuard™ 18 Gauge Needle
Use these needles in the Thermo Scientific™ Dionex™ OnGuard™ Workstation to connect to Dionex OnGuard II Cartridges and direct the flow of liquid into collection vials.
Have Questions?
Catalog NumberFor Use With (Equipment)
039996OnGuard Sample Preparation Station
Catalog number 039996
Price (USD)
4.66
Each 1
For Use With (Equipment):
OnGuard Sample Preparation Station
Price (USD)
4.66
Each 1
Used on the Thermo Scientific™ Dionex™ OnGuard™ Workstation, Dionex OnGuard 18 Gauge Needles connect to the Leur-lock fitting that attaches to the Dionex OnGuard II Cartridges and direct the flow of liquid into collection vials. Dionex OnGuard II Cartridges remove sample matrix interferences encountered in many ion chromatography (IC) applications. Up to 12 cartridges can be placed on the vacuum-driven Dionex OnGuard™ Workstation for sample pretreatment in parallel.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description18 Gauge, 1.25/Luer
QuantityEach 1
For Use With (Equipment)OnGuard Sample Preparation Station
Type18 Gauge Needle
Unit SizeEach 1