Thermo Scientific Chemicals

Sieve Pans, polished brass, 1in deep

Catalog number: 039999.ON
7.62 cm, Each
Thermo Scientific Chemicals

Sieve Pans, polished brass, 1in deep

Catalog number: 039999.ON
7.62 cm, Each
Diameter (Metric) Outer
Catalog number: 039999.ON
also known as 039999-ON
Price (TWD)
Form
Sieve Pan, polished brass, 1 inch. deep

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text