Thermo Scientific Chemicals

Sodium tetraborate decahydrate, ACS, 99.5-105.0%, Thermo Scientific Chemicals

Catalog number: 040114.36
500 g, Each
Thermo Scientific Chemicals

Sodium tetraborate decahydrate, ACS, 99.5-105.0%, Thermo Scientific Chemicals

Catalog number: 040114.36
500 g, Each
Quantity
Catalog number: 040114.36
also known as 040114-36
Price (USD)
Price: 45.60
Online price: 38.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1303-96-4
IUPAC Name
disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
Molecular Formula
B4H20Na2O17
InChI Key
OJPFLODMGUZMKD-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O
Impurity content
Cl 0.001%
Impurity content
PO₄ 0.001%
Insoluble material
0.005%
Assay (unspecified)
99.5-105.0% B₄Na₂O₇.1₀H₂O
Impurity content
Ca 0.005%

Description

It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text