Sodium metabisulfite, ACS, 97% min
Sodium metabisulfite, ACS, 97% min
Sodium metabisulfite, ACS, 97% min
Thermo Scientific Chemicals

Sodium metabisulfite, ACS, 97% min

CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 040115.36
also known as 040115-36
Price (USD)
38.65
Special offer
Online exclusive
Ends: 30-Jun-2026
45.80
Save 7.15 (16%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 30-Jun-2026
45.80
Save 7.15 (16%)
Each
Chemical Identifiers
CAS7681-57-4
IUPAC Namedisodium (sulfinatooxy)sulfinate
Molecular FormulaNa2O5S2
InChI KeyLDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)OS([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.0% Na₂O₅S₂ min
Impurity contentFe 0.002%
Insoluble material0.005%
Impurity contentS₂O₃ 0.05%
Impurity contentCl 0.05%
View more
Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only