Sodium iodate, 99% min
Sodium iodate, 99% min
Sodium iodate, 99% min
Thermo Scientific Chemicals

Sodium iodate, 99% min

CAS: 7681-55-2 | INaO3 | 197.891 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 040135.22
Price (EUR)
58,90
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
58,90
Each
Chemical Identifiers
CAS7681-55-2
IUPAC Namesodium iodate
Molecular FormulaINaO3
InChI KeyWTCBONOLBHEDIL-UHFFFAOYSA-M
SMILES[Na+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Sodium iodate is used in animal feed, food industry. It is utilized in the manufacture of disinfectants, antiseptics and deodorants. It acts as a significant part in the oxidation of aromatic, aliphatic, and alicyclic thiols. Potassium tetrahydroborate and sodium iodate in acidic aqueous solution react with cadmium to generate chemical vapor, which is useful for atomic fluorescence spectrometry application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only