ProPac™ PA1 HPLC Columns
Thermo Scientific™

ProPac™ PA1 HPLC Columns

Green features
Efficiently analyze and purify hydrophilic anionic proteins and peptides with ProPac™ PA1 LC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
040137250 mm9 mm
03965750 mm4 mm
039658250 mm4 mm
Catalog number 040137
Price (MXN)
-
Length (Metric):
250 mm
Diameter (Metric):
9 mm
Efficiently analyze and purify hydrophilic anionic proteins and peptides with Thermo Scientific™ ProPac™ PA1 LC Columns. These strong anion-exchange columns provide high-resolution separations of proteins with pI values from 3 to 11.

The columns can also resolve some proteins and peptides that are highly cationic and have very high isoelectric points.

  • Good for hydrophilic proteins and peptides
  • Ideal for high-resolution separations of proteins with pI values from 3 to 11
  • Pellicular packing ensures high-efficiency and fast mass transport
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeIon Exchange
Flow Rate0.9 to 1.5 mL/min
For Use WithBioLC Chromatography System
Green FeaturesFSC
Max. Pressure5000 psi (345 bar)
Packing MaterialSpherical, Nonporous Polymer
QuantityEach
Stationary PhaseAlkyl Quaternary Amine
Diameter (Metric)9 mm
Length (Metric)250 mm
MaterialPEEK
Particle Size10 μm
PhaseIon Exchange
Pore SizeNonporous
Product LineProPac
TechniqueBio-LC
TypeAnalytical Column
Unit SizeEach